Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TIMP2 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1D7, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA504043
Description
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintece of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.Specifications
TIMP2 | |
Monoclonal | |
0.87 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P16035, P25785, P30121, Q9TTY1 | |
CSC-21K, DDC8 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
21858, 29543, 403633, 7077 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D7 | |
Unconjugated | |
TIMP2 | |
Liquid | |
Timp2 | |
Full length human recombit protein of human TIMP2 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Mouse, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title