Learn More
TIMP2 Mouse anti-Human, Clone: 3A4, Abnova™
Mouse monoclonal antibody raised against human TIMP2.
Manufacturer:  Abnova Corporation MAB12979
Catalog No. 89-935-011
Add to cart
Description
Description
This gene is a member of the TIMP gene family. The proteins encoded by this gene family are natural inhibitors of the matrix metalloproteinases, a group of peptidases involved in degradation of the extracellular matrix. In addition to an inhibitory role against metalloproteinases, the encoded protein has a unique role among TIMP family members in its ability to directly suppress the proliferation of endothelial cells. As a result, the encoded protein may be critical to the maintenance of tissue homeostasis by suppressing the proliferation of quiescent tissues in response to angiogenic factors, and by inhibiting protease activity in tissues undergoing remodelling of the extracellular matrix. [provided by RefSeq]
Specifications
Specifications
TIMP2
3A4
Mouse monoclonal antibody raised against human TIMP2.
In PBS (0.08% sodium azide).
CSC-21K
Mouse
IgG2a
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
TIMP2
Human TIMP2.
Protein A/G purification
RUO
Primary
7077