Alfa Aesar™ Tin foil, 0.05mm (0.002 in.) thick, 98.8% (metals basis)

Manufacturer: Alfa Aesar™ 41604CL

Quantity 15x300cm

 View more versions of this product

Catalog No. AA41604CL

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5
    MDL Number MFCD00133862
    Chemical Name or Material Tin foil, 0.05mm (0.002 in.) thick

    Specifications

    Physical Form Foil
    Odor Odorless
    Quantity 15x300cm
    MDL Number MFCD00133862
    CAS 7440-31-5
    Weight ≈8.2g/15x15cm
    TSCA TSCA
    Chemical Name or Material Tin foil, 0.05mm (0.002 in.) thick
    Assay 98.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit