Alfa Aesar™ Tin foil, 0.33mm (0.013 in.) thick, 99+% (metals basis)

Manufacturer: Alfa Aesar™ 14636RD

Quantity 150x270mm

 View more versions of this product

Catalog No. AA14636RD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5
    MDL Number MFCD00133862
    Chemical Name or Material Tin foil, 0.33mm (0.013 in.) thick

    Specifications

    TSCA TSCA
    Physical Form Foil
    MDL Number MFCD00133862
    CAS 7440-31-5
    Odor Odorless
    Weight ≈100g/15x27cm
    Quantity 150x270mm
    Chemical Name or Material Tin foil, 0.33mm (0.013 in.) thick
    Assay ≥99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit