Alfa Aesar™ Tin foil, 2.0mm (0.08 in.) thick, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 43236HP

Quantity 100x500mm

 View more versions of this product

Catalog No. AA43236HP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5
    MDL Number MFCD00133862
    Chemical Name or Material Tin foil, 2.0mm (0.08 in.) thick

    Specifications

    Weight ≈36g/50x50mm
    Physical Form Foil
    MDL Number MFCD00133862
    CAS 7440-31-5
    Odor Odorless
    TSCA TSCA
    Quantity 100x500mm
    Chemical Name or Material Tin foil, 2.0mm (0.08 in.) thick
    Assay 99.8%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit