Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin (Granular, 30 mesh and Finer/Certified), Fisher Chemical

Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german
5352426
CHEBI:32990
tin
[Sn]
7440-31-5
Sn
118.71
MFCD00133862
ATJFFYVFTNAWJD-UHFFFAOYSA-N
powder,stannum,metallic,tin, elemental,wang,zinn,flake,tin, metal,silver matt powder,zinn german
5352426
CHEBI:32990
tin
[Sn]
Quantity 500g
Arsenic (As) 2ppm max.
Copper (Cu) 0.002% max.
Density 7.31g/mL
Identification Pass Test
Iron (Fe) 0.005% max.
Lead (Pb) 0.02% max.
Mesh Size 30 Mesh
Packaging Glass Bottle
Formula Weight 118.71
Physical Form Solid
Grade Certified
Total Foreign Metals 0.05% max.
Vapor Pressure 1mmHg at 1492°C
Atomic Weight 118.69
Color Silver
Melting Point 231.9°C
Chemical Name or Material Tin
Show More Show Less