Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin(II) 2-ethylhexanoate, 96%

Click to view available options
Quantity:
25g
100g
301-10-0
C16H30O4Sn
405.122
MFCD00002676
KSBAEPSJVUENNK-UHFFFAOYSA-L
stannous octoate,tin ii 2-ethylhexanoate,tin octoate,tin ii bis 2-ethylhexanoate,tin 2+ bis 2-ethylhexanoate,tin ethylhexanoate,tin 2-ethylhexanoate,hexanoic acid, 2-ethyl-, tin 2+ salt,stannous 2-ethylhexoate,tin ii 2-ethylhexylate
9318
2-ethylhexanoate;tin(2+)
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
301-10-0
C16H30O4Sn
405.122
MFCD00002676
KSBAEPSJVUENNK-UHFFFAOYSA-L
stannous octoate,tin ii 2-ethylhexanoate,tin octoate,tin ii bis 2-ethylhexanoate,tin 2+ bis 2-ethylhexanoate,tin ethylhexanoate,tin 2-ethylhexanoate,hexanoic acid, 2-ethyl-, tin 2+ salt,stannous 2-ethylhexoate,tin ii 2-ethylhexylate
9318
2-ethylhexanoate;tin(2+)
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
Applications
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Odor Characteristic
Flash Point >110°C (230°F)
Quantity 100g
Formula Weight 405.11
Percent Purity 96%
Chemical Name or Material Tin(II) 2-ethylhexanoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MO7870000

TSCA : TSCA