Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin(II) 2-ethylhexanoate, 96%

Click to view available options
Quantity:
25g
100g
301-10-0
C16H30O4Sn
405.122
MFCD00002676
KSBAEPSJVUENNK-UHFFFAOYSA-L
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
9318
2-ethylhexanoate;tin(2+)
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
301-10-0
C16H30O4Sn
405.122
MFCD00002676
KSBAEPSJVUENNK-UHFFFAOYSA-L
stannous octoate, tin ii 2-ethylhexanoate, tin octoate, tin ii bis 2-ethylhexanoate, tin 2+ bis 2-ethylhexanoate, tin ethylhexanoate, tin 2-ethylhexanoate, hexanoic acid, 2-ethyl-, tin 2+ salt, stannous 2-ethylhexoate, tin ii 2-ethylhexylate
9318
2-ethylhexanoate;tin(2+)
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Sn+2]
Applications
Tin(II) 2-ethylhexanoate is used as a polymerization initiator in polylactic acid production. It acts as an intermediate as well as a catalyst for urethane foams, a lubricant, addition agent and a stabilizer for transformer oils.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
Quantity 25g
Odor Characteristic
Flash Point >110°C (230°F)
Refractive Index 1.494
Density 1.251
Solubility Information Miscible with water.
Formula Weight 405.11
Percent Purity 96%
Chemical Name or Material Tin(II) 2-ethylhexanoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c

H317-H318-H361

EINECSNumber : 206-108-6

RTECSNumber : MO7870000

TSCA : Yes

Recommended Storage : Ambient temperatures