Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin(II) 2-ethylhexanoate, tech.

Manufacturer:  Alfa Aesar™ 1859022

Quantity 100g

 View more versions of this product

Catalog No. AA1859022


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

301-10-0
MFCD00002676
Tin(II) 2-ethylhexanoate
405.11
2-Ethylhexanoic acid tin(II) salt; Stannous octoate
Technical Grade

Specifications

301-10-0
>110° (230°F)
Colorless to pale yellow
Liquid
MFCD00002676
Tin(II) 2-ethylhexanoate
100g
1.251g/mL
1.4940
405.11
2-Ethylhexanoic acid tin(II) salt; Stannous octoate
Technical Grade

Safety and Handling

GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MO7870000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit