Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) chloride, 98%, anhydrous, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
7772-99-8
Cl2Sn
189.6
MFCD00011241
AXZWODMDQAVCJE-UHFFFAOYSA-L
Stannous chloride
24479
CHEBI:78067
dichlorotin
Cl[Sn]Cl
7772-99-8
Cl2Sn
189.6
MFCD00011241
AXZWODMDQAVCJE-UHFFFAOYSA-L
Stannous chloride
24479
CHEBI:78067
dichlorotin
Cl[Sn]Cl
Assay Percent Range 97.5 to 102.5% (Iodimetry)
Linear Formula SnCl2
Fieser 01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334
Merck Index 15, 8910
Solubility Information Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
Formula Weight 189.6
Physical Form Fine Crystalline Powder or Flakes
Percent Purity 98%
Density 3.9500g/mL
Iron (Fe) 50ppm max.
Lead (Pb) 75ppm max.
Packaging Glass bottle
Solubility Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
Specific Gravity 3.95
Boiling Point 652.0°C
Color White
Melting Point 246.0°C
Quantity 2.5kg
Chemical Name or Material Tin(II) chloride, 98%
Show More Show Less

GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause dam

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 231-868-0

RTECSNumber : XP8700000

TSCA : TSCA