Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) chloride dihydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
10025-69-1
Cl2Sn·2H2O
225.63
MFCD00149863
Stannous chloride dihydrate
10025-69-1
Cl2Sn·2H2O
225.63
MFCD00149863
Stannous chloride dihydrate
Assay Percent Range 96% min. (Iodimetry)
Linear Formula SnCl2·2H2O
Fieser 01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68
Merck Index 15, 8910
Solubility Information Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl,soluble in tartaric acid,alcohol,ethyl acetate,and glacial acetic acid
Formula Weight 225.63
Physical Form Adhering Crystals or Crystalline Powder
Percent Purity 97%
Density 2.7100g/mL
Iron (Fe) 50ppm max.
Lead (Pb) 100ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Boiling Point 652.0°C
Color White or Colorless
Melting Point 37.0°C to 38.0°C
Quantity 2.5kg
Chemical Name or Material Tin(II) chloride dihydrate, 97%
Show More Show Less

GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

RTECSNumber : XP8850000