Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 652°C |
Melting Point | 37°C to 38°C |
Color | White or Colorless |
Physical Form | Adhering Crystalline Powder or Crystals |
Quantity | 500 g |
Assay Percent Range | 96% min. (Iodimetry) |
Linear Formula | SnCl2·2H2O |
Fieser | 01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68 |
Merck Index | 15, 8910 |
Solubility Information | Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl,soluble in tartaric acid,alcohol,ethyl acetate,and glacial acetic acid |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May be corrosive to metals. Harmful if inhaled. Causes severe skin burns and eye damage. May cause an allergic skin reaction. May cause respiratory irritation. May cause damage to organs through prolon |
Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER |
RTECSNumber | XP8850000 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction