Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(II) chloride dihydrate, low in Mercury, suitable for AAS, Acros Organics

Click to view available options
Quantity:
50g
250g
10025-69-1
Cl2Sn·2H2O
225.63
Stannous chloride dihydrate
10025-69-1
Cl2Sn·2H2O
225.63
Stannous chloride dihydrate
Linear Formula SnCl2·2H2O
Ammonium 0.002% max
Arsenic (As) 0.0001% max.
Solubility Information Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Formula Weight 225.63
Percent Purity ≥97.5% (Iodometry)
Calcium (Ca) 0.005% max.
Copper (Cu) 0.001 ù max.
Insoluble Matter 0.005% max
Iron (Fe) 0.002% max.
Lead (Pb) 0.005% max.
Magnesium (Mg) 0.005% max.
Manganese (Mn) 0.0005% max.
Mercury (Hg) 0.000001% max.
Nickel (Ni) 0.0005% max.
Potassium (K) 0.005% max.
Sodium (Na) 0.005% max.
Solubility Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
Sulfate 0.002% max
Boiling Point 652.0°C
Melting Point 37.0°C to 38.0°C
Quantity 250g
Chemical Name or Material Tin(II) chloride dihydrate
Show More Show Less

GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger