Tin(II) chloride dihydrate, Reagent Grade , Alfa Aesar™

Manufacturer: Alfa Aesar 1153609

Quantity 10g

 View more versions of this product

Catalog No. AA1153609

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 10025-69-1
    Physical Form Crystalline
    Molecular Formula SnCl2·2H2O
    Formula Weight 225.65 (189.62 Anhydrous)
    MDL Number MFCD00149863
    Synonym Stannous chloride dihydrate
    Chemical Name or Material Tin(II) chloride dihydrate, Reagent Grade

    Specifications

    Formula Weight 225.65 (189.62 Anhydrous)
    Quantity 10g
    EINECS Number 231-868-0
    Physical Form Crystalline
    Molecular Formula SnCl2·2H2O
    MDL Number MFCD00149863
    CAS 10025-69-1
    Synonym Stannous chloride dihydrate
    Chemical Name or Material Tin(II) chloride dihydrate, Reagent Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit