Learn More
Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Description
Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 43°C to 46°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Linear Formula | SnCl2·xH2O |
UN Number | UN3260 |
Merck Index | 14,8783 |
Solubility Information | Soluble in ethanol,acetone,ether and tetrahydrofuran. Insoluble in xylene. |
IUPAC Name | dichlorotin |
Formula Weight | 189.62 (Anhydrous) |
Show More |
Safety and Handling
Hazard Category | H290-H302+H332-H314-H317-H335-H373 |
Hazard Statement | GHS H Statement H314-H318-H302-H317 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 231-868-0 |
RTECSNumber | XP8850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.