Tin(II) Chloride, Ultra Dry, 99.9985% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 3572314

Quantity 25g

 View more versions of this product

Catalog No. AA3572314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7772-99-8
    Physical Form Powder
    Assay Percent Range 99.9%, metal basis
    Molecular Formula SnCl2
    Formula Weight 189.62
    MDL Number MFCD00011241
    Synonym Stannous chloride
    Chemical Name or Material Tin(II) Chloride, Ampouled under argon
    Grade Ultra dry

    Specifications

    Formula Weight 189.62
    Quantity 25g
    DOT Information DOT Class: 8
    Health Hazard 2 Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    EINECS Number 231-868-0
    Physical Form Powder
    Sensitivity Air, moisture sensitive
    Assay Percent Range 99.9%, metal basis
    Molecular Formula SnCl2
    MDL Number MFCD00011241
    CAS 7772-99-8
    Synonym Stannous chloride
    Health Hazard 1 Harmful if swallowed. Causes burns. May cause sensitization by skin contact.
    Chemical Name or Material Tin(II) Chloride, Ampouled under argon
    Grade Ultra dry

    Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    Harmful if swallowed. Causes burns. May cause sensitization by skin contact.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit