Alfa Aesar™ Tin(II) fluoride, 97.5%

Manufacturer: Alfa Aesar™ 1154422

Quantity 100g

 View more versions of this product

Catalog No. AA1154422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-47-3
    Linear Formula SnF2
    Formula Weight 156.69
    MDL Number MFCD00042540
    UN Number UN3260
    Merck Index 14,8784
    Chemical Name or Material Tin(II) fluoride

    Specifications

    Formula Weight 156.69
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD00042540
    CAS 7783-47-3
    Boiling Point 850°C
    Color White to off-white
    Melting Point 219°C
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    TSCA TSCA
    UN Number UN3260
    Merck Index 14,8784
    Quantity 100g
    Density 4.57g/mL
    Odor Odorless
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    RTECS Number XQ3450000
    Linear Formula SnF2
    Chemical Name or Material Tin(II) fluoride
    Assay 97.5%

    GHS H Statement
    H314-H318-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit