Tin(II) Pyrophosphate, Alfa Aesar™

Manufacturer: Alfa Aesar 8978518

Quantity 50g

 View more versions of this product

Catalog No. AA8978518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15578-26-4
    Physical Form Powder
    Molecular Formula Sn2P2O7
    Formula Weight 411.32
    MDL Number MFCD00049544
    Synonym Tin(II) phosphate
    Chemical Name or Material Tin(II) Pyrophosphate

    Specifications

    Formula Weight 411.32
    Quantity 50g
    EINECS Number 239-635-5
    Physical Form Powder
    Density 4.009
    Molecular Formula Sn2P2O7
    MDL Number MFCD00049544
    CAS 15578-26-4
    Synonym Tin(II) phosphate
    Health Hazard 1 Irritating to eyes, respiratory system and skin.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
    Chemical Name or Material Tin(II) Pyrophosphate

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

    Irritating to eyes, respiratory system and skin.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit