Learn More
Tin(II) trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals
Description
Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Quantity | 1 g |
UN Number | UN3288 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 416.82 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | Tin(II) trifluoromethanesulfonate |
Safety and Handling
Hazard Category | H314-H500 |
Hazard Statement | GHS H Statement H331-H302-H312 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.