Alfa Aesar™ Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis)

Manufacturer: Alfa Aesar™ 3916914

Quantity 25g

 View more versions of this product

Catalog No. AA3916914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 288374-32-3
    Formula Weight 523.08
    MDL Number MFCD00210604
    Synonym Tin(IV) 2-ethylhexano-isopropoxide
    Chemical Name or Material Tin(IV) 2-ethylhexanoate diisopropoxide

    Specifications

    Formula Weight 523.08
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Physical Form Liquid
    MDL Number MFCD00210604
    Synonym Tin(IV) 2-ethylhexano-isopropoxide
    Health Hazard 1 Warning
    Sensitivity Moisture Sensitive
    CAS 288374-32-3
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H361-H315-H319
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material Tin(IV) 2-ethylhexanoate diisopropoxide
    Assay 98%, (metals basis)

    GHS H Statement
    H361-H315-H319
    Suspected of damaging fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit