Tin(IV) Acetate, Alfa Aesar™

Manufacturer: Alfa Aesar 8992106

Quantity 5g

 View more versions of this product

Catalog No. AA8992106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2800-96-6
    Physical Form Crystalline
    Molecular Formula Sn(OOCCH3)4
    Formula Weight 354.87
    MDL Number MFCD00014976
    Chemical Name or Material Tin(IV) Acetate

    Specifications

    Formula Weight 354.87
    Quantity 5g
    Physical Form Crystalline
    Molecular Formula Sn(OOCCH3)4
    MDL Number MFCD00014976
    CAS 2800-96-6
    Chemical Name or Material Tin(IV) Acetate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit