Alfa Aesar™ Tin(IV) chloride, 1M soln. in dichloromethane

Manufacturer: Alfa Aesar™ H32379AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH32379AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7646-78-8
    Molecular Formula Cl4Sn
    Linear Formula SnCl4
    Formula Weight 260.5
    MDL Number MFCD00011242
    UN Number UN2922
    Synonym Stannic chloride
    Chemical Name or Material Tin(IV) chloride

    Specifications

    Formula Weight 260.5
    TSCA TSCA
    Linear Formula SnCl4
    CAS 7646-78-8
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Molecular Formula Cl4Sn
    MDL Number MFCD00011242
    UN Number UN2922
    Synonym Stannic chloride
    Quantity 500mL
    Health Hazard 2 GHS H Statement
    H314-H318-H351
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.
    Concentration 1M soln. in dichloromethane
    Chemical Name or Material Tin(IV) chloride

    GHS H Statement
    H314-H318-H351
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing cancer.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit