Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) chloride, 99.999%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
7646-78-8
Cl4Sn
260.52
MFCD00011242
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4
24287
tetrachlorostannane
Cl[Sn](Cl)(Cl)Cl
7646-78-8
Cl4Sn
260.52
MFCD00011242
HPGGPRDJHPYFRM-UHFFFAOYSA-J
tin tetrachloride,tin iv chloride,stannane, tetrachloro,tin chloride,stannic chloride,tetrachlorotin,tin perchloride,tintetrachloride,tin iv tetrachloride,sncl4
24287
tetrachlorostannane
Cl[Sn](Cl)(Cl)Cl
For ultra trace analysis, Lewis acid
Assay Percent Range 99.999%
Linear Formula SnCl4
Fieser 01,1111; 03,269; 05,627; 06,553; 07,342; 09,436; 11,522; 12,486; 13,14; 14,304; 15,311; 17,335
Merck Index 15, 8901
Solubility Information Solubility in water: reacts. Other solubilities: soluble in carbon disulfide,alcohol,carbon,tetrachloride,benzene,toluene,acetone,kerosene,and gasoline
Formula Weight 260.52
Physical Form Liquid
Percent Purity 99.999%
Grade Trace Metal Basis
Density 2.2200g/mL
Packaging Glass bottle
Solubility Solubility in water: reacts. Other solubilities: soluble in carbon disulfide, alcohol, carbon, tetrachloride, benzene, toluene, acetone, kerosene, and gasoline
Specific Gravity 2.22
Total Trace Metal Impurities 0.001% max
Boiling Point 114.0°C
Color Colorless
Melting Point -33.0°C
Quantity 50g
Chemical Name or Material Tin(IV) chloride, 99.999%
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 231-588-9