Alfa Aesar™ Tin(IV) dibromide bis(2,4-pentanedionate), 98%

Manufacturer: Alfa Aesar™ H5553206

Quantity 5g

 View more versions of this product

Catalog No. AAH5553206

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 16894-10-3
    Formula Weight 476.72
    MDL Number MFCD00075325
    UN Number UN3261
    Synonym Tin(IV) bis(acetylacetonate) dibromide
    Chemical Name or Material Tin(IV) dibromide bis(2,4-pentanedionate)

    Specifications

    Formula Weight 476.72
    Melting Point 182° to 184°C
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container according to local regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    MDL Number MFCD00075325
    Color White to yellow
    Physical Form Powder
    UN Number UN3261
    Synonym Tin(IV) bis(acetylacetonate) dibromide
    Quantity 5g
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    CAS 16894-10-3
    Chemical Name or Material Tin(IV) dibromide bis(2,4-pentanedionate)
    Assay 98%

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container according to local regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit