Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Tin(IV) dibromide bis(2,4-pentanedionate), 98%

Manufacturer:  Alfa Aesar™ H5553214

Quantity 25g

 View more versions of this product

Catalog No. AAH5553214

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

16894-10-3
MFCD00075325
Tin(IV) bis(acetylacetonate) dibromide
476.72
UN3261
Tin(IV) dibromide bis(2,4-pentanedionate)

Specifications

16894-10-3
476.72
UN3261
White to yellow
25g
98%
Powder
MFCD00075325
Tin(IV) bis(acetylacetonate) dibromide
182° to 184°C
Tin(IV) dibromide bis(2,4-pentanedionate)

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container according to local regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit