Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) iodide, 99+%, -10 mesh, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7790-47-8
I4Sn
626.31
MFCD00036272
QPBYLOWPSRZOFX-UHFFFAOYSA-J
stannic iodide,stannane, tetraiodo,tin iv iodide,tetraiodotin,tin tetraiodide,tin iodide,unii-8h688bw69x,tin iv iodide 1:4,tin-iv-iodide
24631
tetraiodostannane
[Sn](I)(I)(I)I
7790-47-8
I4Sn
626.31
MFCD00036272
QPBYLOWPSRZOFX-UHFFFAOYSA-J
stannic iodide,stannane, tetraiodo,tin iv iodide,tetraiodotin,tin tetraiodide,tin iodide,unii-8h688bw69x,tin iv iodide 1:4,tin-iv-iodide
24631
tetraiodostannane
[Sn](I)(I)(I)I
Lewis acid
Assay Percent Range 99+%
Linear Formula SnI4
Merck Index 15, 8904
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,benzene,chloroform,ether and,carbon disulfide
Formula Weight 626.31
Physical Form Powder
Percent Purity ≥99.0%
Packaging Glass bottle
Boiling Point 364°C
Color Brown
Melting Point 144°C
Quantity 10g
Chemical Name or Material Tin(IV) iodide
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger