Alfa Aesar™ Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol

Manufacturer: Alfa Aesar™ 45480AP

Quantity 500mL

 View more versions of this product

Catalog No. AA45480AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1184-61-8
    Molecular Formula C12H28O4Sn
    Linear Formula Sn[OCH(CH3)2]4
    Formula Weight 355.06
    MDL Number MFCD00145408
    UN Number UN3274
    Chemical Name or Material Tin (IV) isopropoxide

    Specifications

    Formula Weight 355.06
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H314-H302-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    Concentration 10% w/v in isopropanol
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
    Sensitivity Moisture Sensitive
    Molecular Formula C12H28O4Sn
    Linear Formula Sn[OCH(CH3)2]4
    MDL Number MFCD00145408
    Color Colorless
    Physical Form Liquid
    CAS 1184-61-8
    Quantity 500mL
    Health Hazard 1 Danger
    UN Number UN3274
    Odor Alcohol-like
    Chemical Name or Material Tin (IV) isopropoxide
    Assay 99%, (metals basis)

    GHS H Statement
    H225-H314-H302-H336
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit