Tin(IV) oxide, 99.996% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 1089114

Quantity 25g

 View more versions of this product

Catalog No. AA1089114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18282-10-5
    Physical Form -22 Mesh Powder
    Assay Percent Range 99.996% (metals basis)
    Molecular Formula O2Sn
    Formula Weight 150.69
    MDL Number MFCD00011244
    Synonym Stannic oxide
    Chemical Name or Material Tin(IV) oxide

    Specifications

    Formula Weight 150.69
    Quantity 25g
    EINECS Number 242-159-0
    Physical Form -22 Mesh Powder
    Assay Percent Range 99.996% (metals basis)
    Molecular Formula O2Sn
    MDL Number MFCD00011244
    CAS 18282-10-5
    Synonym Stannic oxide
    Chemical Name or Material Tin(IV) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit