Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tin(IV) Oxide, Extra Pure, MilliporeSigma™

Click to view available options
Quantity:
250g
1kg
5kg
25kg
Packaging:
Fibre carton
Plastic bottle
18282-10-5
O2Sn
150.708
XOLBLPGZBRYERU-UHFFFAOYSA-N
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
29011
CHEBI:52991
dioxotin
O=[Sn]=O
18282-10-5
O2Sn
150.708
XOLBLPGZBRYERU-UHFFFAOYSA-N
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
29011
CHEBI:52991
dioxotin
O=[Sn]=O
Appearance Off-white solid
Formula Weight 150.69. (molar mass)
Physical Form Solid
Grade Extra Pure
Odor Odorless
Packaging Plastic bottle
Quantity 5kg
Chemical Name or Material Tin(IV) oxide