Alfa Aesar™ Tin(IV) tert-pentyloxide, 5% w/v in 2-methyl-2-butanol, 99% (metals basis)

Manufacturer: Alfa Aesar™ 36562AE

Quantity 100mL

 View more versions of this product

Catalog No. AA36562AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 467.27
    MDL Number MFCD00145409
    UN Number UN3274
    Chemical Name or Material Tin(IV) tert-pentyloxide

    Specifications

    Formula Weight 467.27
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H314-H318-H302-H332
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.
    Boiling Point 100°C (0.2mmHg)
    MDL Number MFCD00145409
    UN Number UN3274
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
    Sensitivity Moisture Sensitive
    Physical Form Liquid
    Quantity 100mL
    Concentration 5% w/v in 2-methyl-2-butanol
    Chemical Name or Material Tin(IV) tert-pentyloxide
    Assay 99%, (metals basis)

    GHS H Statement
    H225-H314-H318-H302-H332
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit