Alfa Aesar™ Tin plating powder, electroless, part A

Manufacturer: Alfa Aesar™ 44176P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA44176P2

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD03703330
    UN Number UN1759
    Chemical Name or Material Tin plating powder, Electroless, part A

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD03703330
    UN Number UN1759
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H351-H361-H302-H317
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Color Various
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Quantity 1 lb.
    Chemical Name or Material Tin plating powder, Electroless, part A

    Description

    • To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection

    GHS H Statement
    H314-H351-H361-H302-H317
    Causes severe skin burns and eye damage.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit