Tin shot, 99.999% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1101022

Quantity 100g

 View more versions of this product

Catalog No. AA1101022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5
    Physical Form Shot, 3mm (0.1 in.)
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00133862
    Chemical Name or Material Tin

    Specifications

    Quantity 100g
    Color Silver-gray
    Density 7.31g/cm3
    EINECS Number 231-141-8
    Physical Form Shot, 3mm (0.1 in.)
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00133862
    CAS 7440-31-5
    Odor Odorless
    Appearance Silver Gray shot
    Chemical Name or Material Tin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit