Alfa Aesar™ Tin slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 4341622

Quantity 100g

 View more versions of this product

Catalog No. AA4341622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-31-5
    Formula Weight 118.710g/mol
    MDL Number MFCD00133862
    Chemical Name or Material Tin slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
    Grade Puratronic™

    Specifications

    Density 7.310g/mL
    Odor Odorless
    Melting Point 231.9°C
    Boiling Point 2270 deg C
    Quantity 100g
    Weight ≈1.5g/slug
    CAS 7440-31-5
    Physical Form Slug
    Formula Weight 118.710g/mol
    MDL Number MFCD00133862
    TSCA TSCA
    Chemical Name or Material Tin slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length
    Assay 99.995%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit