Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TINAG Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 197471AP

Catalog No. 50-173-2523


Add to cart

Description

Description

The antibody is specific to TINAG.

Mediates adhesion of proximal tubule epithelial cells via integrins alpha3-beta1 and alphaV-beta3. This is a non catalytic peptidase C1 family protein.
Specifications

Specifications

TINAG
0.49 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UJW2
TIN AG, TINAG
Rabbit
IgG
150 μL
-20°C
Primary
26944, 27283
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TINAG
Liquid
Tinag
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit