Tingley™ Better Grade PVC Work Boots

Blend of high-quality PVC resins, low-temperature plasticizers, and nitrile rubber creates work boots that remain supple even in cold weather

Manufacturer: Tingley Rubber Co. 51244.09

 View more versions of this product

Catalog No. 19-022952

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Ideal for agriculture, industrial, and food processing applications.
    Certifications/Compliance ANSI Z41 PT99 M/I/75/C/75.
    Height (English) 15 in.
    Height (Metric) 38cm
    Color Brown
    Material PVC; Steel toe
    Size 9
    ANSI Standard ANSI Z41 PT99 M/I/75/C/75.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit