Learn More
Tingley™ Better Grade PVC Work Boots
Blend of high-quality PVC resins, low-temperature plasticizers, and nitrile rubber creates work boots that remain supple even in cold weather
Manufacturer:  Tingley Rubber Co. 51244.06
 View more versions of this product
Catalog No. 19-022948
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PVC; Steel toe
ANSI Z41 PT99 M/I/75/C/75.
15 in.
Brown
6
Ideal for agriculture, industrial, and food processing applications.
38cm