Learn More
Tingley™ Eagle™ Polyurethane on Nylon Suits: Overalls
Breathable polyurethane on nylon garments for all-day comfort and wear
$45.15
Specifications
Color Yellow
Material Nylon/Polyurethane
Suspender Premium snap-lock
View More Specs
Products 3
Catalog Number Mfr. No. Size Size (English) Waist Size (Metric) Waist Price Quantity & Availability  
Catalog Number Mfr. No. Size Size (English) Waist Size (Metric) Waist Price Quantity & Availability  
19-022762
View Documents
Tingley™
O21007.LG
Large 40 to 42 in. 101.6 to 106.6cm Each for $45.15
Add to cart
19-022764
View Documents
Tingley™
O21007.MD
Medium 36 to 38 in. 91.4 to 96.5cm Each for $45.15
Add to cart
19-022763
View Documents
Tingley™
O21007.XL
X-Large 44 to 46 in. 111.7 to 116.8cm Each for $45.15
Add to cart
Description
Description
Tingley's polyurethane on nylon suits are tough and generously cut to fit over the heaviest of work clothes.
  • Made of 200 denier nylon
  • Interior polyurethane coating and exterior durable water repellency coating for 100% waterproof protection
  • Mildew resistant
  • Resistant to most hydrocarbon oils, organic acids, salts, alkalies and some organic solvents
Transportation, meat packing, utilities, municipalities and general industry.
Compliance
Breathable as per test method ASTM E96
Specifications
Specifications
Yellow
Nylon/Polyurethane
Premium snap-lock
Not recommended in ketones, alcohols and certain solvents.