Learn More
Tingley™ Icon™ Premium Outerwear
Engineered to provide superior comfort and safety in the toughest work environments
Manufacturer:  Tingley™ O24122.MD
 View more versions of this product
Catalog No. 19-166-1155
Add to cart
Description
Description
For use in environments where high visibility is desired.
  • Constructed of breathable, 12mil thick polyurethane on 300 denier woven polyester
  • Stitched and taped sealed seams provide 100% waterproof protection
  • 2 in. (5cm) reflective tape reflects for 360° nighttime conspicuity
  • Overhead shoulder design for complete freedom of movement
  • Mesh/taffeta lining provides added comfort
  • Velcro™ take-up straps on sleeves and ankle cuffs provide adjustable fit
  • Zipper/storm fly front with throat guards seals out wind and rain
  • Full vented cape back offers improved circulation
  • Roll-away hood in collar design
  • D-ring access for fall protection harness
  • With hand warmer side pockets, front cargo pockets with Velcro closure, front radio pocket, and inner breast pocket with zipper closure
  • Fluorescent yellow/green
  • Jacket: attached hood and tapered sweep with adjustable elastic draw cord
  • Bib overalls: heavy-duty suspenders with quick-release buckles and crossover, take up snaps on waist for adjustable fit, and snap-fly front
Compliance
Meets ANSI™/ISEA 107-2010 standards; Class 3 compliant. Overalls are Class E compliant.
Specifications
Specifications
Snap Fly Front
ANSI ISEA 107 Class E, Level 2
300 Denier Woven Polyester
Hook and loop take-up straps at the leg cuffs, suspenders, Plastic suspender divider and quick release buckles
12mil
Tape sealed
116.8cm
29 in.
Polyurethane
Fluorescent Yellow, Green
Medium
300 Denier Polyester
2 in. Silver reflective stripes
46 in.
73.6cm