Learn More
Tingley™ Iron Eagle™ Polyurethane Rainsuit
Extremely comfortable and flexible
Manufacturer:  Tingley™ O22007.SM
 View more versions of this product
Catalog No. 19-059-344A
Add to cart
Description
Description
Lightweight and durable. Resists fats, oils, pine tars, gasoline, grease and a variety of other chemicals.
  • Snag and abrasion resistant
  • Heat sealed seams
  • Batwing sleeve construction with a generous cut
  • Therm-O-Rad™ sealed seams
  • Storm fly front
  • Premium snap-lock suspender buckles
  • Heavy-duty suspenders with crossover
  • Heavy-duty snaps
  • Mildew resistant
Order Info
Overalls available in sizes Small to 6X and jacket in sizes Small to 4X. Contact your Fisher Safety Customer Service Representative for ordering information on sizes not shown.
Specifications
Specifications
Safety
32 to 34 in.
71.1cm
Small
Snap-lock suspender
28 in.