Tingley™ Iron Eagle™ Polyurethane Rainsuit

Extremely comfortable and flexible

Manufacturer: Tingley Rubber Co. O22007.LG

 View more versions of this product

Catalog No. 19-059-344C

  Check Availability
Add to Cart

    Specifications

    Size (English) Waist 40 to 42 in.
    Suspender Snap-lock suspender
    Length (Metric) Inseam 76.2cm
    Length (English) Inseam 30 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit