Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Neoprene Snugleg Work Boots

Superb fit with reinforced uppers

$29.78 - $92.00

Specifications

Certifications/Compliance Exceeds ASTM™ F2413-05 M/I/75/C/75.
Color Red, Beige
Material Neoprene
For Use With (Application) Excellent for use in jobs involving solvents, caustics, hydrocarbons, oils, fats and acids.
Height (English) 12.5 in.
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-022736
View Documents
Tingley™
MB924B.08
8
Pair for $92.00
Add to cart
 
19-022737
View Documents
Tingley™
MB924B.09/DEL
9
Pair for $92.00
Add to cart
 
19-022730
View Documents
Tingley™
MB924B.10
10
Pair for $92.00
Add to cart
 
19-022733
View Documents
Tingley™
MB924B.13/DEL
13
Pair for $92.00
Add to cart
 
19-022732
View Documents
Tingley™
MB924B.12
12
Pair for $29.78

Save 50% Reg : $59.57

Add to cart
 
19-022731
View Documents
Tingley™
MB924B.11
11
Pair for $92.00
Add to cart
 
Description

Description

Excellent for use in jobs involving solvents, caustics, hydrocarbons, oils, fats and acids.

  • Triple dipped with two neoprene coatings
  • Sealant ensures that foot is surrounded by a layer of waterproof and chemical-resistant neoprene for optimal protection
  • Safety-Loc™ outsole and removable polyurethane contour insole for maximum comfort
  • 12.5 in. high (32cm)
  • Neoprene work boots are EH rated
  • Brick-red uppers, brown soles

Compliance

Steel toe exceeds ASTM™ F2413-05 M/I/75/C/75.

Specifications

Specifications

Exceeds ASTM™ F2413-05 M/I/75/C/75.
Neoprene
12.5 in.
Red, Beige
Excellent for use in jobs involving solvents, caustics, hydrocarbons, oils, fats and acids.
32 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit