Tingley™ Neoprene Snugleg Work Boots

Superb fit with reinforced uppers

Manufacturer: Tingley Rubber Co. MB924B.07

 View more versions of this product

Catalog No. 19-022735

  Check Availability
Add to Cart

    Specifications

    Height (Metric) 32cm
    Color Red with Beige Soles
    Material Neoprene
    Size 7
    For Use With (Application) Excellent for use in jobs involving solvents, caustics, hydrocarbons, oils, fats and acids.
    Certifications/Compliance Exceeds ASTM™ F2413-05 M/I/75/C/75.
    Height (English) 12.5 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit