Tingley™ Premier™ Work Boots

Provide excellent slip performance on surfaces

Manufacturer: Tingley Rubber Co. 93245.14

 View more versions of this product

Catalog No. 19-034-462J

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM
    For Use With (Application) Excellent choice for food processing and petrochemical applications
    Height (English) 15 in.
    Sole Features Removable contour cushion insoles, Chevron-Plus outsole
    Toe Material Steel
    Waterproof Yes
    Color Brick Red, Cream
    Material PVC
    Size 14

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit