Tingley™ Safetyflex Flame Resistant Coverall

Manufacturer: Tingley Rubber Co. V41108.XL

 View more versions of this product

Catalog No. 19-150-1307

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASTM D6413
    Material PVC, Polyester
    Seam Type Therm-O-Rad™ sealed
    Color Green
    Size X-Large
    For Use With (Application) Heavy exterior coating for excellent permeation performance
    Collar Type High
    Resistance Flame, industrial acids, chemical and caustics
    Closure Type Storm fly front

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit