Learn More
Tingley™ Snugleg Boots: Neoprene, Steel Toe
Every boot is dipped with a neoprene coating and sealant, so the foot is surrounded by a layer of 100% waterproof and chemical-resistant neoprene for protection and comfort
Manufacturer:  Tingley Rubber Co. MB922B.11X
 View more versions of this product
Catalog No. 19-022727
  Check Availability
Add to cart
Disclaimers:
Chemical Resistance: Animal fats and blood, oils, certain acids, alcohols, alkalies, and certain solvents. Electrical Hazard soles and heels are intended to reduce the hazards due to accidental contact with live electrical circuits, electrically energized conductors, parts or apparatus. Electric Hazard soles and heels are not intended for wear in those work environments where volatile chemicals or explosives may be present, where conductive footwear is required.
Specifications Description & Specifications
Specifications
Neoprene Rubber
ASTM F2413 M/I/75/C/75
12.5 in.
Steel
Steel
11
Designed for tough industrial environments
Fabric
Chevron outsole, Polyurethane contour cushion insole
Brick Red, Brown