Tingley™ Snugleg Boots: Neoprene, Steel Toe

Every boot is dipped with a neoprene coating and sealant, so the foot is surrounded by a layer of 100% waterproof and chemical-resistant neoprene for protection and comfort

Manufacturer: Tingley Rubber Co. MB922B.12X

 View more versions of this product

Catalog No. 19-022728

  Check Availability
Add to Cart

Disclaimers:

Chemical Resistance: Animal fats and blood, oils, certain acids, alcohols, alkalies, and certain solvents. Electrical Hazard soles and heels are intended to reduce the hazards due to accidental contact with live electrical circuits, electrically energized conductors, parts or apparatus. Electric Hazard soles and heels are not intended for wear in those work environments where volatile chemicals or explosives may be present, where conductive footwear is required.


    Specifications

    Liner Material Fabric
    Shank Material Steel
    Sole Features Chevron outsole, Polyurethane contour cushion insole
    Toe Material Steel
    Color Brick Red, Brown
    Material Neoprene Rubber
    Size 12
    Certifications/Compliance ASTM F2413 M/I/75/C/75
    For Use With (Application) Designed for tough industrial environments
    Height (English) 12.5 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit