Learn More
Tingley™ Vision™ Safety Outerwear
Provides all-season comfort
Manufacturer:  Tingley™ P23122.LG
 View more versions of this product
Catalog No. 19-166-1172
Add to cart
Description
Description
Innovative outerwear is ANSI™ 107 compliant for high visibility and breathability.
  • Constructed of 7mil thick polyurethane on 150 denier woven polyester
  • Stitched and taped sealed seams provide 100% waterproof protection
  • Designed for use in multipurpose applications
  • 2 in. (5.1cm) silver reflective tape reflects light for 360° nighttime conspicuity
  • Full cut shoulder with 90° sleeve-to-body angle promotes maximum freedom of movement
  • Velcro™ take-up straps on sleeves and ankle cuffs provide adjustable fit
  • Storm fly front seals out wind and rain
  • Easy access cargo pockets with Velcro closure
  • Roll-away adjustable hood in collar design
  • Fluorescent yellow/green background material for excellent daytime visibility
  • Side openings on pants with flap closure
  • Mildew resistant
Order Info
Overalls in sizes Small to 5X available. Contact your Fisher Safety Customer Service Representative for ordering information.
Compliance
Jacket meets ANSI™/ISEA 107-2010 standards; Class 3 compliant. Pants are Class E compliant.
Specifications
Specifications
Snap Fly Front
ANSI ISEA 107 Class E, Level 2
150 Denier Woven Polyester
Hook and loop take-up straps at the leg cuffs, elastic waist with drawcord
7mil
Tape sealed
48 in.
76.2cm
Polyurethane
Fluorescent Yellow, Green
Large
150 Denier Polyester
2 in. Silver reflective stripes
2.0
121.9cm
30 in.