Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tinidazole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Fiber Drum
19387-91-8
C8H13N3O4S
247.27
HJLSLZFTEKNLFI-UHFFFAOYSA-N
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O
19387-91-8
C8H13N3O4S
247.27
HJLSLZFTEKNLFI-UHFFFAOYSA-N
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O

T1352, 19387-91-8, C8H13N3O4S

Spectrum™ Tinidazole, USP is an antibiotic that is used to treat certain types of vaginal infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 25 g
CAS Max % 1
Percent Purity 98 to 102%
Grade USP