Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tiotropium bromide

Potent muscarinic receptor antagonist

$185.00 - $779.00

Specifications

Chemical Name or Material (1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide
Molecular Formula C19H22BrNO4S2
CAS 136310-93-5
Product Type Target: Non-selective mAChR Antagonists
Formula Weight 472.42
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
590210 View Documents Tocris Bioscience™
5902/10
10mg Each for $185.00
Add to cart
 
590250 View Documents Tocris Bioscience™
5902/50
50mg Each for $779.00
Add to cart
 
Description

Description

Potent muscarinic receptor antagonist. Inhibits electrical field stimulation-induced contraction of guinea pig trachea in vitro (IC50 = 0.17 nM). Induces smooth muscle relaxation and bronchodilation.
Specifications

Specifications

(1α,2β,4β,5α,7β)-7-[(2-Hydroxy-2,2-di-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide
136310-93-5
472.42
C19H22BrNO4S2
Target: Non-selective mAChR Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit