Learn More
Andwin Scientific TIP NEPTUNE FINN 10UL PK960
SDP
Supplier:  Andwin Scientific 89140930
Encompass_Preferred
The 10 L Barrier Pipet Tips by Neptune are specially designed with high density polyethlene filters to avoid sample contamination and with S3 polymer tips to help remove sample hold up. It is sold in a pack of 960 tips.
Catalog No. 50-112-6054